Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
SphI

SphI

SphI, 10000 units/ml, 500 units
Catalog # CA101228-946
Supplier #:  R0182S
SphI
Catalog # CA101228-946
CAS Number:  
SphI
SphI

SphI, 10000 units/ml, 500 units

CAS Number:  
Price
117,01 $
Log in pour la disponibilité
Each
Minimum order quantity is 1
Product Details & Documents

An E.coli strain that carries the SphI gene from Streptomyces phaeochromogenes (NRRL B-3559).

Spécifications

Spécifications

  • Concentration:
    10000 units/ml
  • Désignation:
    SphI
  • Taille:
    500 units
  • Cat. No.:
    CA101228-946
  • Enzyme name:
    SphI

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.