PvuII
PvuII
PvuII, 50000 units/ml, 5000 units
Catalog # CA101228-828
Supplier #:  R0151T
PvuII
Catalog # CA101228-828
CAS Number:  
PvuII
PvuII
PvuII, 50000 units/ml, 5000 units
CAS Number:  
Price
$110.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
An E.coli strain that carries the PvuII gene from Proteus vulgaris (ATCC 13315).
Specifications
Specifications
  • Concentration:
    50000 units/ml
  • Désignation:
    PvuII
  • Taille:
    5000 units
  • Cat. No.:
    CA101228-828
  • Enzyme name:
    PvuII
Specifications