MboI
MboI, 5000 units/ml, 500 units
Catalog # CA101228-804
 :  R0147S
MboI
Catalog # CA101228-804
CAS Number:  
MboI
MboI
MboI, 5000 units/ml, 500 units
CAS Number:  
Price
$127.99
Log in pour la disponibilité
Each
Product Details & Documents
An E.coli strain that carries the MboI gene from Moraxella bovis (ATCC 10900).
Spécifications
Spécifications
  • Concentration:
    5000 units/ml
  • Désignation:
    MboI
  • Taille:
    500 units
  • Cat. No.:
    CA101228-804
  • Enzyme name:
    MboI
Spécifications