Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
MboI

MboI

MboI, 5000 units/ml, 2500 units
Catalog # CA101228-800
Supplier #:  R0147L
MboI
Catalog # CA101228-800
CAS Number:  
MboI
MboI

MboI, 5000 units/ml, 2500 units

CAS Number:  
Price
$533.99
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

An E.coli strain that carries the MboI gene from Moraxella bovis (ATCC 10900).

Specifications

Specifications

  • Concentration:
    5000 units/ml
  • Désignation:
    MboI
  • Taille:
    2500 units
  • Cat. No.:
    CA101228-800
  • Enzyme name:
    MboI

Specifications