Lambda DNA, New England Biolabs
Lambda DNA, 250 µg
Catalog # CA101228-432
 :  N3011S
Lambda DNA, New England Biolabs
Catalog # CA101228-432
CAS Number:  
Lambda DNA, New England Biolabs
Lambda DNA, New England Biolabs
Lambda DNA, 250 µg
CAS Number:  
Price
$124.00
Log in for availability
Each
Product Details & Documents
Duplex DNA is isolated from bacteriophage lambda (cI857ind 1 Sam 7)
Lambda DNA is 48,502 base pairs in length. The phage is isolated from the heat-inducible lysogen E. coli l cI857 S7. The DNA is isolated from the purified phage by phenol extraction and dialyzed against 10 mM Tris-HCl (pH 8.0), 1 mM EDTA.
Specifications
Specifications
  • Désignation:
    Lambda DNA
  • Taille:
    250 µg
Specifications