Cuvette Stirring Rods, FireflySci
Cuvette stirring rod with flared end for macro cuvettes
  76016-684
Marketsource
 :  P67
Cuvette Stirring Rods, FireflySci
  76016-684
Marketsource
Cuvette Stirring Rods, FireflySci
Cuvette Stirring Rods, FireflySci
Cuvette stirring rod with flared end for macro cuvettes
$87.99
Each
Stirring rods for the manual stirring of samples in a 10 mm cuvette.
  • Steam autoclavable at 121 °C (250 °F)
Stirring rods allow samples to be continuously spun at the bottom of a cuvette to prevent sedimentation and coagulation.
  • Désignation:
    Cuvette stirring rod with flared end for macro cuvettes