Screw Caps, Firefly
Open Screw Cap with PTFE Septa, 13-425, Black
76016-674
: SC2
Caps are made of polypropylene and are equipped with a poly-vinyl liner.
Ideal for use with mild acids, alkalis, solvents, alcohols, oils, and aqueous products.
- Color:Black
- Désignation:Open Screw Cap with PTFE Septa, 13-425