Screw Caps, Firefly
Open Screw Cap with PTFE Septa, 13-425, Black
  76016-674
 :  SC2
Screw Caps, Firefly
  76016-674
Screw Caps, Firefly
Screw Caps, Firefly
Open Screw Cap with PTFE Septa, 13-425, Black
$39.41
Each
Caps are made of polypropylene and are equipped with a poly-vinyl liner.
Ideal for use with mild acids, alkalis, solvents, alcohols, oils, and aqueous products.
  • Color:
    Black
  • Désignation:
    Open Screw Cap with PTFE Septa, 13-425