Screw Caps, Firefly | Avantor
Screw Caps, Firefly
Closed Screw Cap with PTFE Septa, 13-425, Black
Catalog # 76016-670
Supplier:   FIREFLYSCI, INC.
Fournisseur #:  SC1
Screw Caps, Firefly
Catalog # 76016-670
Supplier:  FIREFLYSCI, INC.
Screw Caps, Firefly
Screw Caps, Firefly
Closed Screw Cap with PTFE Septa, 13-425, Black
Price
$39.41
Log in pour la disponibilité
Each
Product Details & Documents
Caps are made of polypropylene and are equipped with a poly-vinyl liner.
Ideal for use with mild acids, alkalis, solvents, alcohols, oils, and aqueous products.
Spécifications
  • Color:
    Black
  • Désignation:
    Closed Screw Cap with PTFE Septa, 13-425