Screw Caps, Firefly
Closed Screw Cap with PTFE Septa, 13-425, Black
Catalog # 76016-670
Supplier: FIREFLYSCI, INC.
Fournisseur #: SC1
Jump to:
Product Details & Documents
Caps are made of polypropylene and are equipped with a poly-vinyl liner.
Ideal for use with mild acids, alkalis, solvents, alcohols, oils, and aqueous products.
Spécifications
- Color:Black
- Désignation:Closed Screw Cap with PTFE Septa, 13-425