Disposable Cuvettes for Fluorometer, FireflySci
Disposable cuvette, type 9FLP, semi-micro, 1.500 µl, L×W 10×4 mm, 12.5 mm, 45 mm, 12.5 mm, 10 mm, Polystyrene
Catalog # 76016-592
Supplier: FIREFLYSCI, INC.
Fournisseur #: 9FLPMMA
Jump to:
Product Details & Documents
These cuvettes are used for rapid assays and comes in different materials polystyrene and acrylic.
- Four windows and base optically polished
- With open top
Spécifications
- Volume:1.500 µl
- Désignation:Disposable cuvette, type 9FLP, semi-micro
- Matière:Polystyrene
- Lightpath:10 mm
- Internal Dimensions:L×W 10×4 mm
- Outer Depth:12.5 mm
- Outer Height:45 mm
- Outer Width:12.5 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



