Cuvette Washer, FireflySci | Avantor
Cuvette Washer, FireflySci
Double Cuvette Washer Kit
Catalog # 76015-790
Supplier:   FIREFLYSCI, INC.
Fournisseur #:  P65DKIT
Cuvette Washer, FireflySci
Catalog # 76015-790
Supplier:  FIREFLYSCI, INC.
Cuvette Washer, FireflySci
Cuvette Washer, FireflySci
Double Cuvette Washer Kit
Price
$1,001.25
Log in pour la disponibilité
Each
Product Details & Documents
Cuvette washers are handmade and built to last for multiple uses without contamination.
  • Fast cuvette cleaning action
  • Leave cuvettes in place for quick drying action
  • Use with cuvettes from 3 to10mm lightpath
  • Reduces handling and risk of breakage
Easy to use device requires the cuvette to be turned upside down when the cleaning solution is run.
Ordering information: Washers includes Gasket and stopper. Vacuum flask and pump must be ordered separately. Double cuvette washer kit includes cuvette washer head, rubber gasket, stopper (size 7), filtering flask, and vinyl tubing (304.8 cm). Kits are also available with vacuum pump.
Spécifications
  • Désignation:
    Double Cuvette Washer Kit
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Small Rubber Gasket for Single & Double Cuvette Washer
    Small Rubber Gasket for Single & Double Cuvette Washer
    76015-792
    Log in pour la disponibilité
    Each - $38.13
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Size 7 Vacuum Flask Stopper for Cuvette Washers
    Size 7 Vacuum Flask Stopper for Cuvette Washers
    76015-794
    Log in pour la disponibilité
    Each - $83.74