Cuvette Washer, FireflySci
Single Cuvette Washer Kit
Catalog # 76015-788
Supplier: FIREFLYSCI, INC.
Fournisseur #: P65SKIT
Jump to:
Product Details & Documents
Cuvette washers are handmade and built to last for multiple uses without contamination.
- Fast cuvette cleaning action
- Leave cuvettes in place for quick drying action
- Use with cuvettes from 3 to10mm lightpath
- Reduces handling and risk of breakage
Easy to use device requires the cuvette to be turned upside down when the cleaning solution is run.
Ordering information: Washers includes Gasket and stopper. Vacuum flask and pump must be ordered separately. Double cuvette washer kit includes cuvette washer head, rubber gasket, stopper (size 7), filtering flask, and vinyl tubing (304.8 cm). Kits are also available with vacuum pump.
Spécifications
- Désignation:Single Cuvette Washer Kit
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Small Rubber Gasket for Single & Double Cuvette WasherSmall Rubber Gasket for Single & Double Cuvette Washer76015-792Log in pour la disponibilitéEach - $38.13
Informations sur le produit
- Catalog #AvailabilityPrice
- Size 7 Vacuum Flask Stopper for Cuvette WashersSize 7 Vacuum Flask Stopper for Cuvette Washers76015-794Log in pour la disponibilitéEach - $83.74