Ultraviolet Fluorometer Cuvettes, FireFlySci
10 mm, Quartz cap
Catalog # 76015-380
Supplier:   FIREFLYSCI, INC.
 :  11FLUV10
Ultraviolet Fluorometer Cuvettes, FireFlySci
Catalog # 76015-380
Supplier:  FIREFLYSCI, INC.
Ultraviolet Fluorometer Cuvettes, FireFlySci
Ultraviolet Fluorometer Cuvettes, FireFlySci
10 mm, Quartz cap
Price
$1,096.40
Log in pour la disponibilité
Each
Product Details & Documents
Ultraviolet Fluorometer Cuvettes have four windows with base polished and quartz cap.
Spécifications
  • Cap Type:
    Quartz cap
  • Lightpath:
    10 mm