Ultraviolet Fluorometer Cuvettes, FireFlySci
5 mm, Quartz cap
Catalog # 76015-378
Marketsource
Supplier: FIREFLYSCI, INC.
Fournisseur #: 11FLUV5
Ultraviolet Fluorometer Cuvettes, FireFlySci
Catalog # 76015-378
Supplier: FIREFLYSCI, INC.
Marketsource
Jump to:
Product Details & Documents
Ultraviolet Fluorometer Cuvettes have four windows with base polished and quartz cap.
Spécifications
- Cap Type:Quartz cap
- Lightpath:5 mm