Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Macro Cuvettes, Type 11, FireFlySci

0.70 ml, 2 mm, Glass cap
Catalog # 76015-358
Marketsource
Supplier:   FIREFLYSCI, INC.
 :  11IR2
Macro Cuvettes, Type 11, FireFlySci
Catalog # 76015-358
Supplier:  FIREFLYSCI, INC.
Marketsource
Macro Cuvettes, Type 11, FireFlySci
Macro Cuvettes, Type 11, FireFlySci

0.70 ml, 2 mm, Glass cap

Price
1 260,68 $
Log in pour la disponibilité
Each
 
Product Details & Documents

FireflySci's Type 11 cuvette comes standard with a glass cap, which provides an airtight seal, ensured by the ground finish on the inside of the cap and the top of the cuvette.

  • Hand made ensuring the highest quality optical polish with no defects
  • Firefly cuvettes are guaranteed to give high optical transmission
  • Fire fused to ensure the best bonding
  • Several different materials available: (Optical Glass, UV Quartz, IR Quartz)
  • Lightpaths from 1 mm up to 50 mm available
  • Two windows optically polished

The cap is polished, which enables optical transmission.

Every cell is thoroughly inspected before leaving the factory.

 : Includes airtight glass cap.

Spécifications
  • Cap Type:
    Glass cap
  • Volume:
    0.70 ml
  • Lightpath:
    2 mm