Shimadzu Type 16 Multi-Micro Cell Array, FireflySci
UV Quartz, 100 µl (16×), 10 mm
Catalog # 76015-216
Marketsource
Supplier: FIREFLYSCI, INC.
Supplier #: 16UV10
Shimadzu Type 16 Multi-Micro Cell Array, FireflySci
Catalog # 76015-216
Supplier: FIREFLYSCI, INC.
Marketsource
Jump to:
Product Details & Documents
Multi-Micro Cell Array has 16-chambered multi-micro array of cells for doing multiple measurements.
- Highly accurate results, only need to test sample once
- Made by hand, ensuring high quality optical polishing with no defects
- Fire-fused to ensure that cuvettes are robust and durable
- Comes in UV Quartz
- Available in 10 mm lightpath
- Two windows optically polished
- Walls are made from black quartz to block out stray light
Typically used in Shimadzu machines.
Each cell goes through a rigorous inspection process before leaving the factory.
Specifications
- Volume:100 µl (16×)
- Désignation:UV Quartz
- Lightpath:10 mm