Storage Bins
Shelf bin set (Six SOVE3015-6 bins), Blue, PP, Pack of 6, 11.81×5.20×3.94"
Catalog # 76015-066
Supplier: Treston
Fournisseur #: SBS6-3015-6
Jump to:
Product Details & Documents
These high quality, durable Treston bins have been designed for daily use in stores, production facilities and warehouses.
- Straight sides ensure an efficient use of space
- Multiple color options make finding the right item easy
- Open front provides easy access
- Corrugated base helps retrieve contents, especially smaller sized and flat objects
Ordering information: Labels and dividers sold separately, (translucent plastic), labels and blanking piece FP-6M to produce flat surface for adhesive barcode labels on inside rear wall of bins. Contact Avantor representative for more information. Installation is required. Assembly videos available.
Spécifications
- Color:Blue
- Désignation:Shelf bin set (Six SOVE3015-6 bins)
- Dimensions:11.81×5.20×3.94"
- Matière:PP
- Taille:Pack of 6
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



