VWR® Round Bottom Flask, 3 Necks, Parallel
500 ml, 158 mm, 105 mm, 29/42, 24/40
- Ø ext.:105 mm
- Capacité:500 ml
- Hauteur:158 mm
- Center Neck [ST] Joint:29/42
- Side Neck ST Joints:24/40
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



