LumiLoupe Plus
LumiLoupe Plus, 5.5×
Catalog # 470227-870
Supplier: CARSON OPTICAL
Fournisseur #: LO-06
Jump to:
Product Details & Documents
A loupe with a transparent base to allow ambient light to illuminate the viewing object.
- Allows for viewing of an object's fine detail
- Dual lenses for close-up viewing and less distortion
Spécifications
- Grossissement:5.5×
- Désignation:LumiLoupe Plus