$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
G.E.M. LINE ADJUSTABLE WRIST STRAP SET
Catalog # 76001-720
Supplier:
BOTRON COMPANY INC.
Expand image
G.E.M. LINE ADJUSTABLE WRIST STRAP SET
Catalog # 76001-720
Supplier:
BOTRON COMPANY INC.
Supplier Number:
B9938
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Universal Pipette Tips
Supplier: VWR International
Acétonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH