Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TEST PAPER UNVERSAL 1-14 DISPNSR 5M ROLL
Catalog # 75996-726
Supplier:
Macherey-Nagel
Certificates
Expand image
TEST PAPER UNVERSAL 1-14 DISPNSR 5M ROLL
Catalog # 75996-726
Supplier:
Macherey-Nagel
Supplier Number:
90204
Certificates
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
Acétonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International