Micro Vessel Scissors, Sklar
All Sklar instruments are industry standard.
- Reusable
- Nonsterile
- Désignation:Micro Vessel Scissors
- Longueur:191 mm (7½")
- Matière:Stainless steel
- Curvature:Curved