Micro Scissors, Sklar
Product Details & Documents
Sklar's® micro scissors are used for the dissection of tissues, blood vessels and nerves.
- Reusable
- Nonsterile
Available with a flat or round handle. The straight scissors may be used to cut sutures and trim nerve endings. Curved will dissect vessels and nerves. In addition to the tips, the sides may be used for dissection with the tips closed, similar to using forceps.
Specifications
- Désignation:Micro Scissors, Round Handle
- Longueur:152 mm (6")
- Matière:Stainless steel
- Curvature:Straight
- Blade Type:Sharp/Sharp, Smooth