Elevator, Sklar | Avantor
Elevator, Sklar
19.1 cm (7½"), Curved
Catalog # 75995-550
Supplier:   Sklar
Fournisseur #:  98-7316
Elevator, Sklar
Catalog # 75995-550
Supplier:  Sklar
Elevator, Sklar
Elevator, Sklar
19.1 cm (7½"), Curved
Price
$562.79
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Multipurpose instrument for use in micro neurosurgical procedures.
  • Reusable
  • Nonsterile
The elevator is designed to elevate tissue layers to gain clear access to explore the surgical field. The elevator features a straight shaft and comes with either a curved or angled tip.
Spécifications
  • Longueur:
    19.1 cm (7½")
  • Curvature:
    Curved