Objectives for REVOLVE Microscopes, Echo
Achromat objective, 4×, 18.5 mm, 0.10 NA
  75992-492
Objectives for REVOLVE Microscopes, Echo
  75992-492
 :  
Objectives for REVOLVE Microscopes, Echo
Achromat objective, 4×, 18.5 mm, 0.10 NA
  • Grossissement:
  • Désignation:
    Achromat objective
  • Aperture:
    0.10 NA
  • Working Distance:
    18.5 mm