Objectives for REVOLVE Microscopes, Echo
Fluorite phase objective, 4×, 17 mm, 0.13 NA
Catalog # 75992-490
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG0043
Jump to:
Product Details & Documents
Spécifications
- Grossissement:4×
- Désignation:Fluorite phase objective
- Aperture:0.13 NA
- Working Distance:17 mm