Objectives for REVOLVE Microscopes, Echo
Fluorite objective, 4×, 17 mm, 0.13 NA
Catalog # 75992-488
Fournisseur #: JMAG0042
Jump to:
Discontinued
Product Details & Documents
Spécifications
- Grossissement:4×
- Désignation:Fluorite objective
- Aperture:0.13 NA
- Working Distance:17 mm