Objectives for REVOLVE Microscopes, Echo
Apochromat objective, 2×, 6.2 mm, 0.08 NA
Catalog # 75992-482
Supplier: DISCOVER ECHO INC.
: JMAG0020
Jump to:
Product Details & Documents
Spécifications
- Grossissement:2×
- Désignation:Apochromat objective
- Aperture:0.08 NA
- Working Distance:6.2 mm