Objectives for REVOLVE Microscopes, Echo
Apochromat objective, 1.25×, 5 mm, 0.04 NA
Catalog # 75992-480
Supplier: DISCOVER ECHO INC.
: JMAG0011
Jump to:
Product Details & Documents
Spécifications
- Grossissement:1.25×
- Désignation:Apochromat objective
- Aperture:0.04 NA
- Working Distance:5 mm