Objectives for REVOLVE Microscopes, Echo
Achromat oil phase objective, 100×, 0.15 mm, 1.25 NA
Catalog # 75990-684
Supplier: DISCOVER ECHO INC.
: JMAG1006
Jump to:
Product Details & Documents
Spécifications
- Grossissement:100×
- Désignation:Achromat oil phase objective
- Aperture:1.25 NA
- Working Distance:0.15 mm