Objectives for REVOLVE Microscopes, Echo
Achromat oil objective, 100×, 0.15 mm, 1.25 NA
Catalog # 75990-682
Supplier: DISCOVER ECHO INC.
Fournisseur #: JMAG1005
Jump to:
Product Details & Documents
Spécifications
- Grossissement:100×
- Désignation:Achromat oil objective
- Aperture:1.25 NA
- Working Distance:0.15 mm