Objectives for REVOLVE Microscopes, Echo
Objectives for REVOLVE Microscopes, Echo
Fluorite ELWD objective, 60×, 1.5 - 2.2 mm, 0.70 NA
  75990-680
Supplier #:  JMAG0604
Objectives for REVOLVE Microscopes, Echo
  75990-680
Objectives for REVOLVE Microscopes, Echo
Fluorite ELWD objective, 60×, 1.5 - 2.2 mm, 0.70 NA
Each
Minimum order quantity is 1
  • Grossissement:
    60×
  • Désignation:
    Fluorite ELWD objective
  • Aperture:
    0.70 NA
  • Working Distance:
    1.5 - 2.2 mm