Objectives for REVOLVE Microscopes, Echo
Fluorite phase objective, 40×, 0.51 mm, 0.75 NA
Catalog # 75990-666
Fournisseur #: JMAG0406
Jump to:
Discontinued
Product Details & Documents
Spécifications
- Grossissement:40×
- Désignation:Fluorite phase objective
- Aperture:0.75 NA
- Working Distance:0.51 mm