Objectives for REVOLVE Microscopes, Echo
Fluorite ELWD phase objective, 40×, 3.0 - 4.2 mm, 0.60 NA
Catalog # 75990-662
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG0404
Jump to:
Product Details & Documents
Specifications
- Grossissement:40×
- Désignation:Fluorite ELWD phase objective
- Aperture:0.60 NA
- Working Distance:3.0 - 4.2 mm