Objectives for REVOLVE Microscopes, Echo
Fluorite ELWD objective, 40×, 2.7 - 4 mm, 0.60 NA
Catalog # 75990-660
Supplier: DISCOVER ECHO INC.
: JMAG0403
Jump to:
Product Details & Documents
Spécifications
- Grossissement:40×
- Désignation:Fluorite ELWD objective
- Aperture:0.60 NA
- Working Distance:2.7 - 4 mm