Objectives for REVOLVE Microscopes, Echo
Achromat ELWD phase objective, 20×, 3.2 mm, 0.40 NA
Catalog # 75990-654
Supplier: DISCOVER ECHO INC.
Fournisseur #: JMAG0209
Jump to:
Product Details & Documents
Spécifications
- Grossissement:20×
- Désignation:Achromat ELWD phase objective
- Aperture:0.40 NA
- Working Distance:3.2 mm