Objectives for REVOLVE Microscopes, Echo
Fluorite phase objective, 20×, 2.1 mm, 0.50 NA
Catalog # 75990-646
: JMAG0205
Jump to:
Discontinued
Product Details & Documents
Spécifications
- Grossissement:20×
- Désignation:Fluorite phase objective
- Aperture:0.50 NA
- Working Distance:2.1 mm