Objectives for REVOLVE Microscopes, Echo
Fluorite ELWD phase objective, 20×, 6.6 - 7.8 mm, 0.45 NA
Catalog # 75990-642
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG0203
Jump to:
Product Details & Documents
Spécifications
- Grossissement:20×
- Désignation:Fluorite ELWD phase objective
- Aperture:0.45 NA
- Working Distance:6.6 - 7.8 mm