Objectives for REVOLVE Microscopes, Echo
Plan fluorite LWD CC, 20×, 6.6 - 7.8 mm, 0.45 NA
  75990-640
 :  JMAG0202
Objectives for REVOLVE Microscopes, Echo
  75990-640
Objectives for REVOLVE Microscopes, Echo
Plan fluorite LWD CC, 20×, 6.6 - 7.8 mm, 0.45 NA
Each
  • Grossissement:
    20×
  • Désignation:
    Plan fluorite LWD CC
  • Aperture:
    0.45 NA
  • Working Distance:
    6.6 - 7.8 mm