Objectives for REVOLVE Microscopes, Echo
Achromat objective, 10×, 10.6 mm, 0.25 NA
Catalog # 75990-632
Supplier: DISCOVER ECHO INC.
Fournisseur #: JMAG0104
Jump to:
Product Details & Documents
Spécifications
- Grossissement:10×
- Désignation:Achromat objective
- Aperture:0.25 NA
- Working Distance:10.6 mm