Objectives for REVOLVE Microscopes, Echo
Fluorite oil phase objective, 100×, 0.2 mm, 1.30 NA
Catalog # 75990-530
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG1004
Jump to:
Product Details & Documents
Spécifications
- Grossissement:100×
- Désignation:Fluorite oil phase objective
- Aperture:1.30 NA
- Working Distance:0.2 mm