Objectives for REVOLVE Microscopes, Echo
Achromat objective, 60×, 0.17 mm, 0.80 NA
Catalog # 75990-522
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG0608
Jump to:
Product Details & Documents
Specifications
- Grossissement:60×
- Désignation:Achromat objective
- Aperture:0.80 NA
- Working Distance:0.17 mm