Objectives for REVOLVE Microscopes, Echo
Fluorite oil objective, 60×, 0.12 mm, 1.25 - 0.65 NA
Catalog # 75990-518
Supplier: DISCOVER ECHO INC.
: JMAG0606
Jump to:
Product Details & Documents
Spécifications
- Grossissement:60×
- Désignation:Fluorite oil objective
- Aperture:1.25 - 0.65 NA
- Working Distance:0.12 mm