Objectives for REVOLVE Microscopes, Echo
Fluorite ELWD Phase objective, 60×, 1.5 - 2.2 mm, 0.70 NA
75990-516
: JMAG0605
- Grossissement:60×
- Désignation:Fluorite ELWD Phase objective
- Aperture:0.70 NA
- Working Distance:1.5 - 2.2 mm