Maintenance Solutions for Electrodes | Avantor
Maintenance Solutions for Electrodes
Cleaning solution thiourea, colorles, odorless, completely miscible in water, 250 ml
Catalog # 75982-120
Supplier:   Mettler Toledo
Fournisseur #:  51350102
Maintenance Solutions for Electrodes
Catalog # 75982-120
Supplier:  Mettler Toledo
CAS Number:  
Maintenance Solutions for Electrodes
Maintenance Solutions for Electrodes
Cleaning solution thiourea, colorles, odorless, completely miscible in water, 250 ml
CAS Number:  
Price
$53.80
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
This solution keeps your electrode ready with the right cleaning, storage and regeneration solution.
 : Includes quality inspection certificate.
 : Packaging type - bottle.
Spécifications
Spécifications
  • Désignation:
    Cleaning solution thiourea, colorles, odorless, completely miscible in water
  • Taille:
    250 ml