VWR® Sharps Container Systems, Red
2 gal., Stackable, 25.4W×17.8D×28.6H cm (10W×7D×11.25"H), Yellow
Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes, and other sharps.
- Puncture-resistant
- Autoclave-safe and incinerator-safe
- Transparent, locking lids
- Needle removal ports
The stackable sharps containers are puncture-resistant and both autoclaves-safe and incinerator-safe. The tortuous path systems restrict hand entry. The extended neck sharps containers feature a funneled vertical drop and extended handles.
- Capacité:2 gal.
- Color:Yellow
- Dimensions:25.4W×17.8D×28.6H cm (10W×7D×11.25"H)
- Style:Stackable



