Wheatstone Bridge
Product Details & Documents
Bridge has broad, heavily plated brass strips mounted on polished hardwood base.
- A resistance wire negligible temperature coefficient is stretched over a meter ruler with millimeter graduations
Terminals, mounted on five plated brass strips, provide means for connecting resistors and a voltage source. These strips are separated by gaps that can be eliminated (as required) by using two further brass strips. A robust “jockey” (moveable contact) with lead is supplied for taking readings on the wire. A sprung clip provides convenient storage for the jockey.
Ordering information: Required but not included is a power supply and a galvanometer.
Spécifications
- Désignation:Jockey Pen
- Dimensions:–