Wheatstone Bridge | Avantor
Wheatstone Bridge
Jockey Pen, –
Catalog # 470226-560
Supplier:   Wards
Fournisseur #:  PH1162
Wheatstone Bridge
Catalog # 470226-560
Supplier:  Wards
Wheatstone Bridge
Wheatstone Bridge
Jockey Pen, –
Price
$25.30
Log in pour la disponibilité
Each
Product Details & Documents
Bridge has broad, heavily plated brass strips mounted on polished hardwood base.
  • A resistance wire negligible temperature coefficient is stretched over a meter ruler with millimeter graduations
Terminals, mounted on five plated brass strips, provide means for connecting resistors and a voltage source. These strips are separated by gaps that can be eliminated (as required) by using two further brass strips. A robust “jockey” (moveable contact) with lead is supplied for taking readings on the wire. A sprung clip provides convenient storage for the jockey.
Ordering information: Required but not included is a power supply and a galvanometer.
Spécifications
  • Désignation:
    Jockey Pen
  • Dimensions:
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.