Wheatstone Bridge
Wheatstone Bridge
Wheatstone Bridge, 1.13×80×50 mm (two gap)
Catalog # 470226-558
Supplier:   Wards
Supplier #:  PH1156A
Wheatstone Bridge
Catalog # 470226-558
Supplier:  Wards
Wheatstone Bridge
Wheatstone Bridge
Wheatstone Bridge, 1.13×80×50 mm (two gap)
Price
$145.05
Log in for availability
Each
Product Details & Documents
Bridge has broad, heavily plated brass strips mounted on polished hardwood base.
  • A resistance wire negligible temperature coefficient is stretched over a meter ruler with millimeter graduations
Terminals, mounted on five plated brass strips, provide means for connecting resistors and a voltage source. These strips are separated by gaps that can be eliminated (as required) by using two further brass strips. A robust “jockey” (moveable contact) with lead is supplied for taking readings on the wire. A sprung clip provides convenient storage for the jockey.
Ordering information: Required but not included is a power supply and a galvanometer.
Specifications
  • Désignation:
    Wheatstone Bridge
  • Dimensions:
    1.13×80×50 mm (two gap)