Wheatstone Bridge
Product Details & Documents
Bridge has broad, heavily plated brass strips mounted on polished hardwood base.
- A resistance wire negligible temperature coefficient is stretched over a meter ruler with millimeter graduations
Terminals, mounted on five plated brass strips, provide means for connecting resistors and a voltage source. These strips are separated by gaps that can be eliminated (as required) by using two further brass strips. A robust “jockey” (moveable contact) with lead is supplied for taking readings on the wire. A sprung clip provides convenient storage for the jockey.
Ordering information: Required but not included is a power supply and a galvanometer.
Specifications
- Désignation:Wheatstone Bridge
- Dimensions:1.13×80×50 mm (two gap)