Test Kits, COD, TNT plus®
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process
- Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
- Barcode helps the instrument to automatically detect and run the correct method
- Barcode allows spectrophotometer to detect reagent lot number and expiry date
- Vials have flat bottom to improve handling
- Vials are color coded by parameter and range
- No reagent blank necessary
TNTplus™ works only with DR 6000™ UV-VIS, DR 3900™ Benchtop, and DR 1900™ Portable spectrophotometers.
- Méthode chimique:Reactor digestion
- Désignation:COD TNTplus™, HR, Hg-Free (digestion required)
- Taille:150 Tubes/pack
- Range:25 - 1000 mg/L COD