Test Kits, COD, TNT plus® | Avantor
Test Kits, COD, TNT plus®
COD TNTplus™, UHR (digestion required), 25 tubes/pack, Reactor digestion, 250 - 15000 mg/L COD
Catalog # 75919-304
Test Kits, COD, TNT plus®
Catalog # 75919-304
Supplier:  
CAS Number:  
Test Kits, COD, TNT plus®
Test Kits, COD, TNT plus®
COD TNTplus™, UHR (digestion required), 25 tubes/pack, Reactor digestion, 250 - 15000 mg/L COD
CAS Number:  
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
COD-TNTPLUS-ULTRA HIGH RANGE Catalog #CA11016-932
Le produit CA11016-932 est un remplacement de 75919-304 et a le même emballage et les mêmes spécifications, mais peut être d'une quantité différente.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process
  • Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
  • Barcode helps the instrument to automatically detect and run the correct method
  • Barcode allows spectrophotometer to detect reagent lot number and expiry date
  • Vials have flat bottom to improve handling
  • Vials are color coded by parameter and range
  • No reagent blank necessary
TNTplus™ works only with DR 6000™ UV-VIS, DR 3900™ Benchtop, and DR 1900™ Portable spectrophotometers.
Spécifications
Spécifications
  • Méthode chimique:
    Reactor digestion
  • Désignation:
    COD TNTplus™, UHR (digestion required)
  • Taille:
    25 tubes/pack
  • Range:
    250 - 15000 mg/L COD