Exam Table Paper
Deluxe grade smooth paper, 18"×225 ft.
Catalog # 470226-548
Supplier: MEDLINE INDUSTRIES, INC.
Fournisseur #: NON24322
Jump to:
Product Details & Documents
Strong, absorbent table paper protects exam tables from dirt and moisture while offering comfort and protection for patients.
- Features a heavier basis weight paper
- All rolls are mechanically metered for exact length
- Individually poly bagged for cleanliness during shipment and storage
Spécifications
- Désignation:Deluxe grade smooth paper
- Dimensions:18"×225 ft.